Learn More
2-Methyl-5-nitrobenzoic acid, 99+%, Thermo Scientific™
Catalog Number p-3736941
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
1975-52-6
C8H7NO4
181.15
MFCD00007371
DJRFJAVPROZZFL-UHFFFAOYSA-N
2-methyl-5-nitrobenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, 2-methyl-5-nitro-benzoic acid, 5-nitro-2-methylbenzoic acid, rarechem al bo 0285, 5-nitro-2-methyl benzoic acid, 2-methyl-5-nitro benzoic acid, pubchem2244
519683
CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O
1975-52-6
C8H7NO4
181.15
MFCD00007371
DJRFJAVPROZZFL-UHFFFAOYSA-N
2-methyl-5-nitrobenzoic acid, 5-nitro-o-toluic acid, benzoic acid, 2-methyl-5-nitro, 2-methyl-5-nitro-benzoic acid, 5-nitro-2-methylbenzoic acid, rarechem al bo 0285, 5-nitro-2-methyl benzoic acid, 2-methyl-5-nitro benzoic acid, pubchem2244
519683
CC1=C(C=C(C=C1)[N+](=O)[O-])C(=O)O
CAS Min % 99.0
CAS Max % 100.0
Melting Point 176°C to 180°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)CO2H
Quantity 1 g
Beilstein 09, 471
Solubility Information Solubility in water: insoluble
Formula Weight 181.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitrobenzoic acid