Learn More
2-Methyl-5-(trifluoromethyl)aniline, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 25449-96-1 |
| Molecular Formula | C8H8F3N |
| MDL Number | MFCD00134580 |
| Quantity | 25 g |
| SMILES | FC(F)(F)C1=CC=C(C)C(N)=C1 |
| Molecular Weight (g/mol) | 175.15 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Methyl-5-(trifluoromethyl)aniline |