Learn More
2-Methyl-9H-fluoren-9-one, TRC
Product Code. 30472865
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472865 50 mg 50mg
30482884 500 mg 500mg
30471788 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30472865 Supplier Toronto Research Chemicals Supplier No. TRCM32179350MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2840-51-9
Molecular Formula C14H10O
Molecular Weight (g/mol) 194.23
IUPAC Name 2-methylfluoren-9-one
SMILES Cc1ccc2c3ccccc3C(=O)c2c1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C14H10O/c1-9-6-7-11-10-4-2-3-5-12(10)14(15)13(11)8-9/h2-8H,1H3
Formula Weight 194.0732
Chemical Name or Material 2-Methyl-9H-fluoren-9-one