missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl Thiophenothiazine, TRC

Product Code. 30468952
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468952 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468952 Supplier Toronto Research Chemicals Supplier No. TRCM3307151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7643-08-5
Molecular Formula C13 H11 N S2
Quantity 1 g
Synonym 2-Methylthiophenothiazine
InChI Formula InChI=1S/C13H11NS2/c1-15-9-6-7-13-11(8-9)14-10-4-2-3-5-12(10)16-13/h2-8,14H,1H3
SMILES CSc1ccc2Sc3ccccc3Nc2c1
IUPAC Name 2-methylsulfanyl-10H-phenothiazine
Molecular Weight (g/mol) 245.36
Formula Weight 245.0333
Chemical Name or Material 2-Methylthiophenothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.