Learn More
2-Methylisothiazolone Hydrochloride, ≥99.9, MP Biomedicals
Product Code. 11411571 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411571 500 mg 500mg
11421571 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11411571 Supplier MP Biomedicals Supplier No. 0215952290
Chemical Identifiers
CAS 26172-54-3
Molecular Formula C4H6ClNOS
Molecular Weight (g/mol) 151.608
MDL Number MFCD06804636
InChI Key SJXPQSRCFCPWQQ-UHFFFAOYSA-N
Synonym 2-Methyl-4-isothiazoline-3-one, MIT
PubChem CID 117746
IUPAC Name 2-methyl-1,2-thiazol-3-one;hydrochloride
SMILES CN1C(=O)C=CS1.Cl
Specifications
Type 2-Methylisothiazolone Hydrochloride
Physical Form Crystalline
Color White
Quantity 500 mg
Assay Percent Range ≥99%
Formula Weight 151.6
Chemical Name or Material 2-Methylisothiazolone Hydrochloride