Learn More
2-Methylisothiazolone Hydrochloride, ≥99.9, MP Biomedicals
Product Code. 11421571 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421571 1 g 1g
11411571 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11421571 Supplier MP Biomedicals Supplier No. 0215952291
Chemical Identifiers
CAS 26172-54-3
Molecular Formula C4H6ClNOS
Molecular Weight (g/mol) 151.608
MDL Number MFCD06804636
InChI Key SJXPQSRCFCPWQQ-UHFFFAOYSA-N
Synonym 2-Methyl-4-isothiazoline-3-one, MIT
PubChem CID 117746
IUPAC Name 2-methyl-1,2-thiazol-3-one;hydrochloride
SMILES CN1C(=O)C=CS1.Cl
Specifications
Type 2-Methylisothiazolone Hydrochloride
Physical Form Crystalline
Color White
Quantity 1 g
Assay Percent Range ≥99%
Formula Weight 151.6
Chemical Name or Material 2-Methylisothiazolone Hydrochloride
compliance-icons compliance-icons
Product Identifier
  • 2-Methylisothiazolone Hydrochloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .