missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Methylpiperazine, 98%

Product Code. p-7027562
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11445857

Brand: Thermo Scientific Alfa Aesar B24405.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-07-9 | C5H12N2 | 100.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-07-9
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00005954
InChI Key JOMNTHCQHJPVAZ-UHFFFAOYSA-N
Synonym piperazine, 2-methyl, 2-methyl-piperazine, +/--2-methylpiperazine, 2methylpiperazine, 2-methylpiperazin, 2-methypiperazine, 2-methylpiperizine, 3-methylpiperazine, 2-methyl piperazine, pubchem8560
PubChem CID 66057
IUPAC Name 2-methylpiperazine
SMILES CC1CNCCN1

Specifications

Melting Point 62°C to 66°C
Boiling Point 155°C to 156°C
Flash Point 65°C (149°F)
Odor Amine-like
Quantity 25 g
UN Number UN2925
Beilstein 383566
Sensitivity Air sensitive; Hygroscopic
Formula Weight 100.17
Percent Purity 98%
Chemical Name or Material (+/-)-2-Methylpiperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (+/-)-2-Methylpiperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H228-Flammable solid.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : (+/-)-2-Methylpiperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.