missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CSN5i-3-NEG

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
CSN5i-3-NEG is the negative control for CSN5i-3 (Cat. No.7089).
Specifications
Specifications
| Chemical Name or Material | 3-(Difluoromethyl)-N-(6-((5 R,6 R)-6-hydroxy-6,7,8,9-tetrahydro-5 H-imidazo[1,5-a]azepin-5-yl)-[1,10-biphenyl]-3-yl)-1-isopropyl-1 H-pyrazole-5-carboxamide |
| Quantity | 2 mg |
| Target | Ubiquitin Ligase (E3) Inhibitors |
| Molecular Formula | C28H29F2N5O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction