missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TheraPure™ 2'-MOE-5mC Phosphoramidite, serum vial bottle

Product Code. p-7180871 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 18084090

Brand: Thermo Scientific™ 27102002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2'-MOE-5mC CEP is a phosphoramidite containing 5' DMT and the nucleoside 5-methyl-cytidine (5mC) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis.

2'-MOE-5mC CEP is a phosphoramidite containing 5' DMT and the nucleoside 5-methyl-cytidine (5mC) with 2'-O-methoxy-ethyl (MOE) protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methoxy-ethyl (MOE) modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:

  • Full chemical name – 5'-O-(4,4'-Dimethoxytrityl)-2'-O-methoxyethyl-5-methyl-N4-benzoyl-cytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula – C50H60N5O10P
  • Base – 5mC
  • Base Protection – Bz
  • Molecular weight – 922.0 g/mol
  • CAS number – 163759-94-2
  • HPLC purity – ≥99%; M-11 Impurity ≤0.01%
  • 31P NMR purity – ≥99%
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type 2in-MOE-5mC CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Cytidine (C)
Reagent Features 2in-MOE modified
Shipping Condition Room Temperature
Purity 99%
Quantity 1 g
Reagent Type Phosphoramidite
Grade TheraPure™
Nucleic Acid Type RNA
Product Line TheraPure
Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.