Learn More
2-Morpholinecarboxylic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Morpholinecarboxylic Acid Ethyl Ester |
| CAS | 135782-25-1 |
| Molecular Formula | C7H13NO3 |
| Quantity | 100 mg |
| Synonym | 2-Morpholinecarboxylic acid','ethyl ester, Ethyl 2-morpholinecarboxylate, 2-Morpholinecarboxylic acid ethyl ester, Ethyl morpholine-2-carboxylate |
| InChI Formula | InChI=1S/C7H13NO3/c1-2-10-7(9)6-5-8-3-4-11-6/h6,8H,2-5H2,1H3 |
| SMILES | O=C(OCC)C1CNCCO1 |
| IUPAC Name | ethyl morpholine-2-carboxylate |
| Molecular Weight (g/mol) | 159.18 |
| Formula Weight | 159.0895 |