Learn More
2-(n-Butoxycarbonylpiperidine)-5-methyl-1,3-thiazol-4-yl)acetic acid, 97%, Thermo Scientific™
Product Code. p-4510236 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10492803 250 mg Amber Glass Bottle 250mg
10533403 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10492803 Supplier Thermo Scientific Maybridge Supplier No. MO07259CB
Request Bulk or Custom Format
Chemical Identifiers
CAS 845885-88-3
Molecular Formula C16H24N2O4S
Molecular Weight (g/mol) 340.438
InChI Key IHHZQULPYDMAOP-UHFFFAOYSA-N
Synonym 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-yl acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-methyl-1,3-thiazol-4-yl acetic acid, 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-yl aceticacid, 2-5-methyl-2-n-tert-butoxycarbonylpiperidine-1,3-thiazol-4-yl acetic acid, 2-5-methyl-2-piperidine-n-boc protected-1,3-thiazol-4-yl acetic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-methyl-1,3-t, 2-2-1-tert-butoxycarbonyl piperidin-4-yl-5-methylthiazol-4-y, 2-1-butoxycarbonylpiperidine-5-methyl-1,3-thiazol-4-yl acetic acid
PubChem CID 2794810
IUPAC Name 2-[5-methyl-2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]-1,3-thiazol-4-yl]acetic acid
SMILES CC1=C(N=C(S1)C2CCN(CC2)C(=O)OC(C)(C)C)CC(=O)O
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 340.44
Percent Purity 97%
Chemical Name or Material {2-[1-(tert-Butoxycarbonyl)piperidin-4-yl]-5-methyl-1, 3-thiazol-4-yl}acetic acid