Learn More
2-(N-Morpholino)Ethane-Sulfonic Acid, Extra Pure, SLR, Fisher Chemical
Product Code. 10000250 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10000250 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10000250 Supplier Fisher Chemical Supplier No. M/5760/48
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 145224-94-8
Melting Point 300°C
Color White
pH 3.5 to 4.1
Packaging Plastic powder jar
Molecular Formula C6H15NO5S
MDL Number 149409
Quantity 100 g
Synonym mes monohydrate, 2-morpholinoethanesulfonic acid hydrate, mes hydrate, mes buffer hydrate, 4-morpholineethanesulfonic acid monohydrate, 4-morpholineethanesulfonic acid, monohydrate, 2-4-morpholinyl ethanesulfonic acid hydrate, mes hydrate buffer, 4-morpholineethanesulfonicacid, hydrate 1:1, mesmonohydrate
InChI Key MIIIXQJBDGSIKL-UHFFFAOYSA-N
SMILES C1COCCN1CCS(=O)(=O)O.O
IUPAC Name 2-morpholin-4-ylethanesulfonic acid;hydrate
Molecular Weight (g/mol) 213.248
PubChem CID 16218417
Formula Weight 213.25g/mol
Physical Form Powder