Learn More
2-n-Propylfuran, 99%
Product Code. 11450297
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450297 5 g 5g
11460297 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11450297 Supplier Thermo Scientific Alfa Aesar Supplier No. B21703.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 4229-91-8 | C7H10O | 110.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4229-91-8
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00047070
InChI Key CPLJMYOQYRCCBY-UHFFFAOYSA-N
Synonym furan, 2-propyl, 2-n-propylfuran, 2-n-propyl furan, furan, propyl, 2-propyl-furan, furan, alpha-propyl, acmc-1ctud, 2-propyl furan, furan, .alpha.-propyl, cpljmyoqyrccby-uhfffaoysa
PubChem CID 77907
IUPAC Name 2-propylfuran
SMILES CCCC1=CC=CO1
Tekniske data
Density 0.891
Boiling Point 110°C to 111°C
Flash Point 24°C (75°F)
Quantity 5 g
UN Number UN3271
Formula Weight 110.16
Percent Purity 99%
Chemical Name or Material 2-n-Propylfuran
RUO – Research Use Only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.