Learn More
2-Nitro-p-anisidine-15N, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Nitro-p-anisidine-15N |
| CAS | 873990-80-8 |
| Molecular Formula | C7H8N15NO3 |
| Quantity | 100 mg |
| Synonym | 1-Amino-2-nitro-4-methoxybenzene-15N, 2-Nitro-4-methoxyaniline-15N, 2-Nitro-4-methoxybenzenamine-15N, 2-Nitro-p-anisidine-15N, 3-Nitro-4-aminoanisole-15N, 4-Amino-1-methoxy-3-nitrobenzene-15N, 4-Amino-3-nitroanisole-15N, 4-Methoxy-2-nitroaniline-15N, 4-Methoxy-2-nitrobenzenamine-15N, 4-Methoxy-2-nitrohbenzenamine-15N, 4-Methoxy-2-nitrophenylamine-15N, 4-Methoxy-o-nitroaniline-15N, Acco Fast Bordeaux GP Salt-15N, Amarthol Fast Bordeaux GP Base-15N, Amarthol Fast Bordeaux GP Salt-15N, Atul Fast Bordeaux GP Base-15N, Azobase NAS-15N, Azoene Fast Bordeaux GP Base-15N, o-Nitro-p-anisidine-15N, p-Methoxy-o-nitroaniline-15N |
| InChI Formula | InChI=1S/C7H8N2O3/c1-12-5-2-3-6(8)7(4-5)9(10)11/h2-4H,8H2,1H3/i9+1 |
| SMILES | COc1ccc(N)c(c1)[15N+](=O)[O-] |
| IUPAC Name | 4-methoxy-2-[oxido(oxo)azaniumyl]aniline |
| Molecular Weight (g/mol) | 169.14 |
| Formula Weight | 169.0505 |