Learn More
2-Octyn-1-ol, 98%
Product Code. 11387495
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11387495
Brand: Thermo Scientific Alfa Aesar L05596.18
Request Bulk or Custom Format
CAS: 20739-58-6 | C8H14O | 126.20 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20739-58-6
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00039542
InChI Key TTWYFVOMGMBZCF-UHFFFAOYSA-N
Synonym 2-octyn-1-ol, 2-octyne-1-ol, pubchem13066, acmc-1clfg, 2-octyn 1-ol, 2-octyn-1-ol 10g
PubChem CID 140750
IUPAC Name oct-2-yn-1-ol
SMILES CCCCCC#CCO
Specifications
Density 0.88
Boiling Point 76°C to 78°C (2 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.456
Quantity 50 g
Beilstein 1744120
Formula Weight 126.2
Percent Purity 98%
Chemical Name or Material 2-Octyn-1-ol