Learn More
2-Oxopiperazine-3,3,5,5,6,6-d6, CDN
Product Code. 30316044
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316044 0.25 g 0.25g
30320844 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316044 Supplier CDN Supplier No. CDND69040.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Piperazinone-3,3,5,5,6,6-D6
CAS 1219803-71-0
Molecular Formula C4 3 H D5 H2 N2 O
Quantity 0.25 g
Synonym 2-Oxopiperazine-3,3,5,5,6,6-d6, 2-Piperazinone-3,3,5,5,6,6-D6, Piperazinone-d6
InChI Formula InChI=1 S/C4H8N2O/c7-4-3-5-1-2-6-4/h5H,1-3H2,(H,6,7)/i1TD,2D2,3D2
SMILES [2 H]C1([2 H])NC([2 H])([3 H])C([2 H])([2 H])NC1=O
IUPAC Name 3,3,5,6,6-pentadeuterio-5-tritiopiperazin-2-one
Molecular Weight (g/mol) 107.158
Formula Weight 107.1033 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity