missing translation for 'onlineSavingsMsg'
Learn More

2-Palmitoyl-rac-glycerol, TRC

Product Code. 30007820
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007820 50 mg 50mg
30010451 25 mg 25mg
30007152 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007820 Supplier Toronto Research Chemicals Supplier No. TRCP15727050MG
 Restricted Item
Estimated Shipment: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 23470-00-0
Molecular Formula C19 H38 O4
Molecular Weight (g/mol) 330.5
Synonym Palmitin, 2-mono- (6CI,7CI,8CI), 2-Hexadecanoyl glycerol, 2-Monohexadecanoylglycerol, 2-Monopalmitin, 2-Monopalmitoyl-sn-glycerol, 2-Monopalmitoylglycerol, 2-Palmitoylglycerol, Glycerol-β-palmitate, Palmitic acid β-monoglyceride, β-Monopalmitin, Hexadecanoic acid, 2-hydroxy-1-(hydroxymethyl)ethyl ester
IUPAC Name 1,3-dihydroxypropan-2-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(CO)CO

Specifikationer

Quantity 50 mg
InChI Formula InChI=1S/C19H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-18(16-20)17-21/h18,20-21H,2-17H2,1H3
Formula Weight 330.277
Percent Purity >95%
Chemical Name or Material 2-glyceryl monopalmitate
Grade HPLC
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.