missing translation for 'onlineSavingsMsg'
Learn More

2-(Perfluorohexyl)ethyl Thiocyanate, TRC

Product Code. 30417289
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417289 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417289 Supplier Toronto Research Chemicals Supplier No. TRCP28633050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Perfluorohexyl)ethyl Thiocyanate
CAS 26650-09-9
Purity Grade Notes GC
Molecular Formula C9H4F13NS
Quantity 50 mg
Synonym 2-Perfluorohexylethanethiocyanate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl Thiocyanate, 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl Ester Thiocyanic Acid
InChI Formula InChI=1S/C9H4F13NS/c10-4(11,1-2-24-3-23)5(12,13)6(14,15)7(16,17)8(18,19)9(20,21)22/h1-2H2
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCSC#N
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl thiocyanate
Molecular Weight (g/mol) 405.18
Formula Weight 404.9857
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.