Learn More
2-Phenyl-2-[(trimethylsilyl)oxy]acetonitrile (90%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Phenyl-2-[(trimethylsilyl)oxy]acetonitrile |
| CAS | 25438-37-3 |
| Molecular Formula | C11H15NOSi |
| Quantity | 25 mg |
| Synonym | 2-Phenyl-2-(trimethylsilyloxy)ethanenitrile, Phenyl(trimethylsiloxy)acetonitrile, Phenyl(trimethylsilyloxy)acetonitrile, α-(Trimethylsilyloxy)phenylacetonitrile, α-Trimethylsiloxybenzyl Cyanide, α-[(Trimethylsilyl)oxy]benzeneacetonitrile, (Cyanophenylmethoxy)trimethylsilane, 2-(Trimethylsilyloxy)-2-phenylacetonitrile |
| InChI Formula | InChI=1S/C11H15NOSi/c1-14(2,3)13-11(9-12)10-7-5-4-6-8-10/h4-8,11H,1-3H3 |
| SMILES | [Si](OC(C1=CC=CC=C1)C#N)(C)(C)C |
| IUPAC Name | 2-phenyl-2-trimethylsilyloxyacetonitrile |
| Molecular Weight (g/mol) | 205.33 |
| Formula Weight | 205.0923 |