Learn More
2-Piperazinyl-4-amino-6,7-dimethoxyquinazoline, TRC
Product Code. 30010388
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010388 1 g 1g
17952125 50 mg 50mg
17923185 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010388 Supplier Toronto Research Chemicals Supplier No. TRCP4805001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60547-97-9
Molecular Formula C14 H19 N5 O2
Molecular Weight (g/mol) 289.33
Synonym 6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine, Terazosin Hydrochloride Dihydrate Imp. C (EP), Doxazosin Mesilate Imp. G (EP), Prazosin Hydrochloride Imp. C (EP), 4-Quinazolinamine, 6,7-dimethoxy-2-(1-piperazinyl)-, 6,7-Dimethoxy-2-(1-piperazinyl)-4-quinazolinamine, 2-(1-Piperazinyl)-4-amino-6,7-dimethoxyquinazoline, 2-(4-Piperazinyl)-4-amino-6,7-dimethoxyquinazoline, 2-Piperazino-6,7-dimethoxy-4-quinazolinamine, 4-Amino-2-(1-piperazinyl)-6,7-dimethoxyquinazoline, 4-Amino-6,7-dimethoxy-2-(1-piperazinyl)quinazoline
IUPAC Name 6,7-dimethoxy-2-piperazin-1-ylquinazolin-4-amine
SMILES COc1cc2nc(nc(N)c2cc1OC)N3CCNCC3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C14H19N5O2/c1-20-11-7-9-10(8-12(11)21-2)17-14(18-13(9)15)19-5-3-16-4-6-19/h7-8,16H,3-6H2,1-2H3,(H2,15,17,18)
Formula Weight 289.1539
Percent Purity >95%
Chemical Name or Material 6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine
Grade HPLC