Learn More
ZVG Zellstoff Vertrieb™ 2 ply Cleaning Cloth
Click to view available options
Dimensions (L x W):
38 x 38 cm
Unit Size:
2 rolls
Specifications
Specifications
| Product Type | Cleaning Cloth |
| Color | Blue |
| Dimensions (L x W) | 38 x 38 cm |
| Format | Roll |
| For Use With (Equipment) | Multiclean |
| Packaging Quantity | 2 Rolls |
| Ply | 2-ply |