missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol 70% by Volume, Pure, Kraft™

Product Code. 10372203
Click to view available options
Quantity:
10 L
Packaging:
Plastic Cannister, Conductive
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10372203

Brand: Kraft 07131.6700

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for use in solvents for liquid chromatography, in antifreeze compositions and in cosmetics.

  • Synonyms: Isopropyl alcohol, Isopropanol, iso-Propanol, Dimethylcarbinol, 2-Hydroxypropane
  • Molecular Formula: C3H8O
  • Molar Mass: 60.10g/mol
  • CAS: 67-63-0
  • EIN ECS No.: 200-661-7

Specifications

pH Neutral
Melting Point -89.5°C
CAS 67-63-0
Color Colorless
Density 0.875g/cm3
Boiling Point 82°C
Quantity 10 L
Flash Point >17°C
Ignition Point 425°C
Packaging Plastic Cannister, Conductive
Physical Form Liquid
Molecular Formula C3H8O
Vapor Pressure 43hPa (at 20°C)
Water 35.3%
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Solubility Information Fully miscible in water
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1g/mol
Percent Purity 70%
Concentration or Composition (by Analyte or Components) 2-Propanol (70% volume), Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.