missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrid-4-ylpyrimidine-5-carboxylic acid, 90%, Thermo Scientific™

Catalog Number p-3586683
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
259807-47-1
C10H7N3O2
201.185
MFCD09863218
GTKJGOOMSODGOU-UHFFFAOYSA-N
2-pyridin-4-yl pyrimidine-5-carboxylic acid, 2-pyrid-4-ylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-4-pyridinyl, 2-4-pyridyl pyrimidine-5-carboxylic acid, 2-4-pyridinyl-5-pyrimidinecarboxylic acid, 2-pyridin-4-yl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl-9ci
22710213
2-pyridin-4-ylpyrimidine-5-carboxylic acid
C1=CN=CC=C1C2=NC=C(C=N2)C(=O)O
This item is not returnable. View return policy
259807-47-1
C10H7N3O2
201.185
MFCD09863218
GTKJGOOMSODGOU-UHFFFAOYSA-N
2-pyridin-4-yl pyrimidine-5-carboxylic acid, 2-pyrid-4-ylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-4-pyridinyl, 2-4-pyridyl pyrimidine-5-carboxylic acid, 2-4-pyridinyl-5-pyrimidinecarboxylic acid, 2-pyridin-4-yl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl-9ci
22710213
2-pyridin-4-ylpyrimidine-5-carboxylic acid
C1=CN=CC=C1C2=NC=C(C=N2)C(=O)O
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.18
Percent Purity 90%
Chemical Name or Material 2-Pyrid-4-ylpyrimidine-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'