missing translation for 'onlineSavingsMsg'
Learn More

2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride (4-Methyldiphenhydramine Hydrochloride), Mikromol™

Product Code. 30051300
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051300

Brand: Mikromol MM0490.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride (4-Methyldiphenhydramine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride (4-Methyldiphenhydramine Hydrochloride)
CAS 4024-34-4
Analyte or Component Names 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride (4-Methyldiphenhydramine Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C18 H23 N O . Cl H
Certifications/Compliance ISO 17025
Synonym Dimenhydrinate Imp. G (EP), Diphenhydramine Hydrochloride Imp. B (EP), Diphenhydramine Imp. B (EP), Orphenadrine USP Related Compound F, Orphenadrine USP RC F, 4-Methyldiphenhydramine Hydrochloride, (RS)-N,N-Dimethyl-2-[(4-methylphenyl)phenylmethoxy]ethanamine Hydrochloride, N,N-Dimethyl-2-[(RS)-(4-methylphenyl)(phenyl)methoxy]ethanamine Hydrochloride, 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine Hydrochloride, Orphenadrine Hydrochloride Imp. F (EP) as Hydrochloride, para-Methylbenzyl Isomer Hydrochloride of Orphenadrine, Dimenhydrinate Imp. G (EP) as Hydrochloride, Diphenhydramine Hydrochloride Imp. B (EP) as Hydrochloride, Orphenadrine Related Compound F as Hydrochloride, Dimenhydrinate Impurity G as Hydrochloride, Diphenhydramine Hydrochloride Impurity B as Hydrochloride, Orphenadrine Hydrochloride Impurity F as Hydrochloride, Diphenhydramine Impurity B as Hydrochloride, Orphenadrine Impurity F as Hydrochloride
InChI Formula InChI=1S/C18H23NO.ClH/c1-15-9-11-17(12-10-15)18(20-14-13-19(2)3)16-7-5-4-6-8-16;/h4-12,18H,13-14H2,1-3H3;1H
SMILES Cl.O(CCN(C)C)C(C=1C=CC=CC1)C2=CC=C(C=C2)C
IUPAC Name N,N-dimethyl-2-[(4-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride
Molecular Weight (g/mol) 305.84
Quantity 25 mg
Formula Weight 305.1546 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.