Learn More
2-((tert-Butoxycarbonyl)amino)-5-chlorobenzoic acid, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 253677-29-1 |
| Molecular Formula | C12H14ClNO4 |
| MDL Number | MFCD02682158 |
| Quantity | 1 g |
| SMILES | O=C(O)C1=CC(Cl)=CC=C1NC(OC(C)(C)C)=O |
| Molecular Weight (g/mol) | 271.70 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-((tert-Butoxycarbonyl)amino)-5-chlorobenzoic acid |