missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butoxycarbonylamino)-4-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 444630010

Additional Details : CAS : 1150561-75-3 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11996051

  • € 203.00 / 1g


Estimated Shipment: 16-08-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

1150561-75-3
C18H28BNO4
333.24
2-(N-BOC-Amino)-4-methylphenylboronic acid pinacol ester

Specifications

2-(tert-Butoxycarbonylamino)-4-methylphenylboronic acid pinacol ester
128°C to 130°C
1150561-75-3
C18H28BNO4
333.24
97%
Glass Bottle
1g
97%
2-(N-BOC-Amino)-4-methylphenylboronic acid pinacol ester
333.24
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-(tert-Butoxycarbonylamino)-4-methylphenylboronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates