Learn More
2-tert-Butyl-1,3-diisopropylisourea, TRC
Specifications
Specifications
| Chemical Name or Material | 2-tert-Butyl-1,3-diisopropylisourea |
| CAS | 71432-55-8 |
| Molecular Formula | C11H24N2O |
| Quantity | 2.5 g |
| Synonym | N,N'-Bis(1-methylethyl)carbamimidic Acid 1,1-Dimethylethyl Ester, 2-tert-Butyl-1,3-diisopropylpseudourea, N,N'-Diisopropyl-O-tert-butylisourea, O-tert-Butyl-N,N'-diisopropylisourea, tert-Butyl N,N'-dipropan-2-ylcarbamimidate |
| InChI Formula | InChI=1S/C11H24N2O/c1-8(2)12-10(13-9(3)4)14-11(5,6)7/h8-9H,1-7H3,(H,12,13) |
| SMILES | CC(C)N=C(NC(C)C)OC(C)(C)C |
| IUPAC Name | tert-butyl N,N'-diisopropylcarbamimidate |
| Molecular Weight (g/mol) | 200.32 |
| Formula Weight | 200.19 |