Learn More
2-Thiomorpholinoisonicotinic acid, 97%, Thermo Scientific™
Product Code. 10356334
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10356334 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10356334 Supplier Thermo Scientific Maybridge Supplier No. CC50501DA
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Specifications
CAS 884507-29-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H12N2O2S
MDL Number MFCD09025860
Quantity 1 g
Synonym 2-thiomorpholinoisonicotinic acid, 2-thiomorpholin-4-yl pyridine-4-carboxylic acid, 4-pyridinecarboxylicacid, 2-4-thiomorpholinyl, 2-1,4-thiazaperhydroin-4-yl pyridine-4-carboxylic acid
InChI Key DDAWRDLRTZFVHD-UHFFFAOYSA-N
SMILES C1CSCCN1C2=NC=CC(=C2)C(=O)O
IUPAC Name 2-thiomorpholin-4-ylpyridine-4-carboxylic acid
Molecular Weight (g/mol) 224.278
PubChem CID 18525848
Formula Weight 224.28
Percent Purity 97%
Chemical Name or Material 2-Thiomorpholinoisonicotinic acid