missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarbonyl Chloride, TRC

Product Code. 30455379
Click to view available options
Quantity:
25 g
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 30455379

Brand: Toronto Research Chemicals TRCT36809025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Thiophenecarbonyl Chloride
CAS 5271-67-0
Molecular Formula C5H3ClOS
Quantity 25 g
Synonym 2-(Chlorocarbonyl)thiophene, 2-(Chloroformyl)thiophene, 2-Thenoyl Chloride, 2-Thienylcarbonyl chloride, 2-Thiophencarbonyl chloride, 2-Thiophenecarboxylic Acid Chloride, 2-Thiophenecarboxylic Chloride, 2-Thiopheneformyl Chloride, 5-Thiophene-2-carbonyl Chloride, Thenoyl Chloride, Thiophen-2-ylcarbonyl Chloride, alpha-Thenoyl Chloride
InChI Formula InChI=1S/C5H3ClOS/c6-5(7)4-2-1-3-8-4/h1-3H
SMILES ClC(=O)c1cccs1
IUPAC Name thiophene-2-carbonyl chloride
Molecular Weight (g/mol) 146.59
Formula Weight 145.9593
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.