Description
2-tier step for MVE 800 series high efficiency freezers and MVE HECO 815P
- 2-tier step for MVE 800 series high-efficiency freezers
- Step-up for 815, 818, or 819 high-efficiency freezer
- The 2-Tier step assembly enables easy access, insertion, and removal of taller rack systems on the HECO/HE 800 Series
Specifications
Specifications
| For Use With (Equipment) | MVE 800 Series High Efficiency Freezers, MVE HECO 815 P |