Learn More
Chart Australia Pty Ltd 2-Tier Step
Product Code. 18816009 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
MVE 800 Series High Efficiency Freezers, MVE HECO 815 P
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
18816009 MVE 800 Series High Efficiency Freezers, MVE HECO 815 P Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18816009 Supplier Chart Australia Pty Ltd Supplier No. 20820733
  • ONLINE EXCLUSIVE PRICE
    € 12384.48
    € 7692.52 / Pack of 1
    Save € 4691.96
    37% Off
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
2-tier step for MVE 800 series high efficiency freezers and MVE HECO 815P
  • 2-tier step for MVE 800 series high-efficiency freezers
  • Step-up for 815, 818, or 819 high-efficiency freezer
  • The 2-Tier step assembly enables easy access, insertion, and removal of taller rack systems on the HECO/HE 800 Series
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MVE 800 Series High Efficiency Freezers, MVE HECO 815 P