missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine dihydrochloride, 95%, ACROS Organics™

Brand:  Acros Organics 454420010

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15741749

  • € 431.00 / 1g


Estimated Shipment
09-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

1311254-51-9
264.08
2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine dihydrochloride, 2-trifluoromethyl-5h,6h,7h,8h-imidazo 1,2-a pyrazine dihydrochloride
2-(trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine;dihydrochloride
C7H8F3N3·2ClH
NWCODVVXJNWZAL-UHFFFAOYSA-N
56845330
C1CN2C=C(N=C2CN1)C(F)(F)F.Cl.Cl

Specifications

2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine dihydrochloride
Glass Bottle
1g
94.0 to max. 106% min. (Argentometry)
2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine dihydrochloride, 2-trifluoromethyl-5h,6h,7h,8h-imidazo 1,2-a pyrazine dihydrochloride
C1CN2C=C(N=C2CN1)C(F)(F)F.Cl.Cl
264.08
264.08
Authentic
223.0°C to 226.0°C
1311254-51-9
C7H8F3N3·2ClH
NWCODVVXJNWZAL-UHFFFAOYSA-N
2-(trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine;dihydrochloride
56845330
95%
Videos
SDS
Documents