missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine dihydrochloride, 95%, Thermo Scientific™

Catalog Number 15751749
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1311254-51-9
C7H8F3N3·2ClH
264.08
NWCODVVXJNWZAL-UHFFFAOYSA-N
2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine dihydrochloride, 2-trifluoromethyl-5h,6h,7h,8h-imidazo 1,2-a pyrazine dihydrochloride
56845330
2-(trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine;dihydrochloride
C1CN2C=C(N=C2CN1)C(F)(F)F.Cl.Cl
This item is not returnable. View return policy
1311254-51-9
C7H8F3N3·2ClH
264.08
NWCODVVXJNWZAL-UHFFFAOYSA-N
2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine dihydrochloride, 2-trifluoromethyl-5h,6h,7h,8h-imidazo 1,2-a pyrazine dihydrochloride
56845330
2-(trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine;dihydrochloride
C1CN2C=C(N=C2CN1)C(F)(F)F.Cl.Cl
Melting Point 223.0°C to 226.0°C
Infrared Spectrum Authentic
Assay Percent Range 94.0 to max. 106% min. (Argentometry)
Quantity 250 mg
Formula Weight 264.08
Percent Purity 95%
Chemical Name or Material 2-(Trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine dihydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'