missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)oxazolo[4,5-b]pyridine, 97%, Alfa Aesar™

Brand:  Alfa Aesar H63931.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15440909

  • € 178.00 / 1g


Estimated Shipment date
from Supplier 02-07-2020
Add to basket

Specifications

Chemical Identifiers

894406-63-4
188.109
2-trifluoromethyl oxazolo 4,5-b pyridine, 2-trifluoromethyl-1,3 oxazolo 4,5-b pyridine, oxazolo 4,5-b pyridine,2-trifluoromethyl, oxazolo 4,5-b pyridine, 2-trifluoromethyl, 2-trifluoromethyl 1,3 oxazolo 4,5-b pyridine
2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
C7H3F3N2O
MJILPMZYOJELAX-UHFFFAOYSA-N
29919200
C1=CC2=C(N=C1)N=C(O2)C(F)(F)F

Specifications

2-(Trifluoromethyl)oxazolo[4,5-b]pyridine
894406-63-4
2-trifluoromethyl oxazolo 4,5-b pyridine, 2-trifluoromethyl-1,3 oxazolo 4,5-b pyridine, oxazolo 4,5-b pyridine,2-trifluoromethyl, oxazolo 4,5-b pyridine, 2-trifluoromethyl, 2-trifluoromethyl 1,3 oxazolo 4,5-b pyridine
C1=CC2=C(N=C1)N=C(O2)C(F)(F)F
188.109
188.11
97%
1g
C7H3F3N2O
MJILPMZYOJELAX-UHFFFAOYSA-N
2-(trifluoromethyl)-[1,3]oxazolo[4,5-b]pyridine
29919200
Solid
Videos
Documents