Learn More
Lenz Laborglasintrumente 2-Way Stopcocks with PFTE Plug
Click to view available options
Plug Bore:
1.5 mm (Cap)
Unit Size:
Each
Description
- With PTFE plug and screwthread retaining device
- Two oblique bores
- Material: borosilicate glass 3.3
- Leakage rate acc. to DIN 12540-1
Specifications
Specifications
| Plug Bore | 1.5 mm (Cap) |
| Material | Borosilicate Glass (3.3) |