Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x100/4MI Swing Out Rotors
Shop All Ohaus Products
Click to view available options
Capacity:
1 x 100 mL
1 x 15 mL
1 x 25 mL
1 x 30 mL
1 x 50 mL (29 mm dia.)
1 x 50 mL (29 mm dia., FA)
1 x 50 mL (29 mm dia.,RB)
1 x 50 mL (34 mm dia.)
1 x 85 mL
10 x 1.5 mL
2 x 15 mL
5 x 4 to 10 mL
7 x 15 mL
7 x 5 to 7 mL (13 mm dia., RB)
7 x 5 to 7 mL (13 mm dia., Vac)
9 x 4.5 mL
9 x 5 to 7 mL
Unit Size:
Pack of 2
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-S4x100/4MI Swing Out Rotors
Specifications
Specifications
| Type | RB |
| Capacity | 7 x 15 mL |
| Max. Tube Diameter | 17 mm |
| For Use With (Equipment) | R-S4x100/4MI Swing Out Rotors |
| Description | Rack, 7x15ml D17mm RB, 2/pk |
| No. of Places | 7 |
| No. per Pack | 2 |