Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Product Code. 15826752 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
1 x 100 mL
1 x 250 mL
1 x 50 mL (29 mm dia. Rim)
1 x 50 mL (34 mm dia. RB)
10 x 1.6 to 7 mL
14 x 5 to 7 mL
2 x 50 mL
3 x 25 mL
3 x 50 mL (29 mm dia. FA)
3 x 50 mL (29 mm dia. RB)
5 x 25 mL
7 x 15 mL (17 mm dia. FA)
7 x 15 mL (17 mm dia. Rim)
8 x 10 mL
9 x 15 mL
Unit Size:
Pack of 2
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15826752 14 x 5 to 7 mL Pack of 2
15836752 10 x 1.6 to 7 mL Pack of 2
15816752 8 x 10 mL Pack of 2
15886742 9 x 15 mL Pack of 2
15896742 7 x 15 mL (17 mm dia. FA) Pack of 2
15806752 7 x 15 mL (17 mm dia. Rim) Pack of 2
15866742 5 x 25 mL Pack of 2
15876742 3 x 25 mL Pack of 2
15856742 1 x 50 mL (29 mm dia. Rim) Pack of 2
15846742 2 x 50 mL Pack of 2
15816742 3 x 50 mL (29 mm dia. RB) Pack of 2
15826742 3 x 50 mL (29 mm dia. FA) Pack of 2
15836742 1 x 50 mL (34 mm dia. RB) Pack of 2
15806742 1 x 100 mL Pack of 2
15896732 1 x 250 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15826752 Supplier Ohaus™ Supplier No. 30314916
Only null left
Add to basket
Add to basket
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Type RB
Capacity 14 x 5 to 7 mL
Max. Tube Diameter 13 mm
For Use With (Equipment) R-S4x250/4MI Swing Out Rotors
Description Rack, 14x5-7ml D13mm RB, 2/pk
No. of Places 14
No. per Pack 2