Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Product Code. 15856742 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
1 x 100 mL
1 x 250 mL
1 x 50 mL (29 mm dia. Rim)
1 x 50 mL (34 mm dia. RB)
10 x 1.6 to 7 mL
14 x 5 to 7 mL
2 x 50 mL
3 x 25 mL
3 x 50 mL (29 mm dia. FA)
3 x 50 mL (29 mm dia. RB)
5 x 25 mL
7 x 15 mL (17 mm dia. FA)
7 x 15 mL (17 mm dia. Rim)
8 x 10 mL
9 x 15 mL
Unit Size:
Pack of 2
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15856742 1 x 50 mL (29 mm dia. Rim) Pack of 2
15826752 14 x 5 to 7 mL Pack of 2
15836752 10 x 1.6 to 7 mL Pack of 2
15816752 8 x 10 mL Pack of 2
15886742 9 x 15 mL Pack of 2
15896742 7 x 15 mL (17 mm dia. FA) Pack of 2
15806752 7 x 15 mL (17 mm dia. Rim) Pack of 2
15866742 5 x 25 mL Pack of 2
15876742 3 x 25 mL Pack of 2
15846742 2 x 50 mL Pack of 2
15816742 3 x 50 mL (29 mm dia. RB) Pack of 2
15826742 3 x 50 mL (29 mm dia. FA) Pack of 2
15836742 1 x 50 mL (34 mm dia. RB) Pack of 2
15806742 1 x 100 mL Pack of 2
15896732 1 x 250 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15856742 Fournisseur Ohaus™ Code fournisseur 30314909
Il en reste null
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
TRUSTED_SUSTAINABILITY
Spécification
Type Rim
Capacity 1 x 50 mL (29 mm dia. Rim)
Max. Tube Diameter 29 mm
For Use With (Equipment) R-S4x250/4MI Swing Out Rotors
Description Rack, 1x50ml D29mm Rim, 2/pk, V3
No. of Places 1
No. per Pack 2