Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ASTM Certificate, Pore Sizes: Under 10 mm
Product Code. p-8003344 Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
1.18 mm
1.40 mm
1.70 mm
2 mm
2.36 mm
2.80 mm
3.35 mm
4 mm
4.75 mm
5.60 mm
6.30 mm
6.70 mm
8 mm
9.50 mm
Unit Size:
Each
Product Code. 11904662
Brand: RETSCH 60.141.001700
Only null left
Specifications
Quantity 1Each
Pore Size 1.70 mm
Type Test Sieve
Certifications/Compliance ASTM E 11
Material Stainless Steel
For Use With (Equipment) Shaker AS 300
Height (Metric) 25.4mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 203.2 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel