Learn More
RETSCH 203.2 dia. x 25.4mmH Stainless Steel Test Sieve, ISO Certificate, Pore Sizes: 10mm and Larger
€ 172.00 - € 177.00
Specifications
Diameter (Metric) Outer | 203.2 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11934512
|
RETSCH
60.140.010000 |
10mm |
€ 172.00
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
11994532
|
RETSCH
60.140.100000 |
100mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11904542
|
RETSCH
60.140.106000 |
106mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11944512
|
RETSCH
60.140.011200 |
11.20mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11914542
|
RETSCH
60.140.112000 |
112mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11954512
|
RETSCH
60.140.012500 |
12.50mm |
€ 172.00
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
11974512
|
RETSCH
60.140.013200 |
13.20mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11924542
|
RETSCH
60.140.125000 |
125mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11964512
|
RETSCH
60.140.014000 |
14mm |
€ 172.00
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
11984512
|
RETSCH
60.140.016000 |
16mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11994512
|
RETSCH
60.140.018000 |
18mm |
€ 172.00
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
11904522
|
RETSCH
60.140.019000 |
19mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11924522
|
RETSCH
60.140.020000 |
20mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11934522
|
RETSCH
60.140.022400 |
22.40mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11944522
|
RETSCH
60.140.026500 |
26.50mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11914522
|
RETSCH
60.140.025000 |
25mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11954522
|
RETSCH
60.140.028000 |
28mm |
€ 172.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11964522
|
RETSCH
60.140.031500 |
31.50mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11974522
|
RETSCH
60.140.035500 |
35.50mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11984522
|
RETSCH
60.140.037500 |
37.50mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11994522
|
RETSCH
60.140.040000 |
40mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11904532
|
RETSCH
60.140.045000 |
45mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11914532
|
RETSCH
60.140.050000 |
50mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11944532
|
RETSCH
60.140.063000 |
63mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11934532
|
RETSCH
60.140.056000 |
56mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11924532
|
RETSCH
60.140.053000 |
53mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11954532
|
RETSCH
60.140.071000 |
71mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11964532
|
RETSCH
60.140.075000 |
75mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11984532
|
RETSCH
60.140.090000 |
90mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11974532
|
RETSCH
60.140.080000 |
80mm |
€ 177.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Specifications
203.2 mm | |
Stainless Steel | |
1Each | |
ISO 3310/1 | |
Shaker AS 300 |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
25.4mm |