Learn More
Invitrogen™ MDH1 Recombinant Rabbit Monoclonal Antibody (24GB1295)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557455
 View more versions of this product
Product Code. 30274920
  • € 198.00 / 20µL
Description
MDH1 Recombinant Monoclonal Antibody for Western Blot, Flow
MDH1 is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MDH1
Recombinant Monoclonal
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O88989, P14152, P40925
MDH1
A synthesized peptide derived from human MDH1 (aa 300-334).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1295
Unconjugated
MDH1
B230377B03Rik; Cytosolic malate dehydrogenase; D17921; Diiodophenylpyruvate reductase; epididymis secretory protein Li 32; HEL-S-32; malate dehydrogenase (EC 1.1.1.37); malate dehydrogenase 1; malate dehydrogenase 1, NAD (soluble); malate dehydrogenase, cytoplasmic; malate dehydrogenase, cytoplasmic; malate dehydrogenase, peroxisomal; malate dehydrogenase, soluble; Malate dehydrogenase-like enzyme; Mdh; Mdh1; MDHA; Mdhl; MDH-s; MDL1; MGC:1375; Mor2; Mor-2; soluble malate dehydrogenase
Rabbit
Affinity chromatography
RUO
17449, 24551, 4190
-20°C