Learn More
Invitrogen™ NR2C2 Recombinant Rabbit Monoclonal Antibody (24GB730)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557519
 View more versions of this product
Product Code. 30275739
  • € 198.00 / 20µL
Description
NR2C2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Members of the nuclear hormone receptor family, such as NR2C2, act as ligand-activated transcription factors. The proteins have an N-terminal transactivation domain, a central DNA-binding domain with 2 zinc fingers, and a ligand-binding domain at the C terminus. The activated receptor/ligand complex is translocated to the nucleus where it binds to hormone response elements of target genes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NR2C2
Recombinant Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P49116, P49117, P55094
NR2C2
A synthesized peptide derived from human NR2C2 / TR4 (aa 500-555).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB730
Unconjugated
NR2C2
mKIAA4145; Mtr2r1; Nr2c2; Nuclear hormone receptor TR4; nuclear receptor subfamily 2 group C member 2; nuclear receptor subfamily 2, group C, member 2; nuclear receptor subfamily 2, group H, member 2; orphan nuclear receptor TAK1; Orphan nuclear receptor TR4; orphan receptor TR4; orphan receptor, TR4; Tak1; testicular nuclear receptor 4; testicular receptor 4; TR2R1; Tr4; TR4 orphan receptor; TR4-NS orphan receptor
Rabbit
Affinity chromatography
RUO
22026, 50659, 7182
-20°C