Learn More
Hamilton™ 21 gauge, Small Hub RN Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
Product Code. 12336449
Click to view available options
Needle Gauge:
21 G
Unit Size:
Pack of 6
Product Code. 12336449
Brand: Hamilton™ 780412
Small Hub Removable (RN) needles can be used with removable (RN)-style syringes that are 100 μL and smaller. Standard needle length is 2 in. / 51 mm. Removable (RN) needles are available in convenient six packs. Custom lengths and point styles are available.
Specification Sheet
  • Gauge – 21 gauge
  • Point Style – 2, 3, or 4
  • Needle Length – Custom Needle Length
  • Autoclavable – Yes
  • Sales Quantity – 6 needles per pack
  • Hub – Small RN
Specifications
Needle Gauge 21 G
Product Type Small Hub RN