Learn More
Hamilton™ 21 gauge, Small Hub RN Needle, custom length (0.375 to 12 in), point style 2, 3, or 4, 6/PK
Click to view available options
Needle Gauge:
21 G
Unit Size:
Pack of 6
Description
Specification Sheet
- Gauge – 21 gauge
- Point Style – 2, 3, or 4
- Needle Length – Custom Needle Length
- Autoclavable – Yes
- Sales Quantity – 6 needles per pack
- Hub – Small RN
Specifications
Specifications
| Needle Gauge | 21 G |
| Product Type | Small Hub RN |