Learn More
23-Keto Nemadectin (Desmethoxyamino Moxidectin), TRC
Product Code. 30006904
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006904 10 mg 10mg
30004240 5 mg 5mg
30006277 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006904 Supplier Toronto Research Chemicals Supplier No. TRCK19553510MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 112124-81-9
Molecular Formula C36H50O8
Molecular Weight (g/mol) 610.78
Synonym (6R,25S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-6,28-epoxy-23-oxomilbemycin B, [6R,25S(E)]-5-O-Demethyl-28-deoxy-25-(1,3-dimethyl-1-butenyl)-6,28-epoxy-23-oxo-milbemycin B, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran] Milbemycin B derivative;
IUPAC Name (2S,2a'E,2a1'S,4'E,5R,6S,6'R,8'E,11'R,15'S,17a'R,20'R,20a'R)-2a1',20'-dihydroxy-5,6',8',19'-tetramethyl-6-((E)-4-methylpent-2-en-2-yl)-2a1',5,6,6',7',10',11',14',15',17a',20',20a'-dodecahydro-2'H,17'H-spiro[pyran-2,13'-[11,15]methano[1,5]dioxacyclooctadecino[9,8,7-cd]benzofuran]-4,17'(3H)-dione
SMILES C/C(C[C@@H](C)/C=C/C=C(CO1)/[C@]2(O)[C@@]1([H])[C@H](O)C(C)=C[C@@]2([H])C3=O)=C\C[C@]4([H])C[C@@](O3)([H])C[C@@]5(O[C@H](/C(C)=C/C(C)C)[C@@H](C)C(C5)=O)O4
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C36H50O8/c1-20(2)13-24(6)32-25(7)30(37)18-35(44-32)17-28-16-27(43-35)12-11-22(4)14-21(3)9-8-10-26-19-41-33-31(38)23(5)15-29(34(39)42-28)36(26,33)40/h8-11,13,15,20-21,25,27-29,31-33,38,40H,12,14,16-19H2,1-7H3/b9-8+,22-11+,24-13+,26-10+/t21-,25-,27+,28-,29-,31+,32+,33+,35-,36+/m0/s1
Formula Weight 610.35
Percent Purity >95%
Chemical Name or Material 23-Keto Nemadectin (Desmethoxyamino Moxidectin)
Grade HPLC