missing translation for 'onlineSavingsMsg'
Learn More

23-Keto Nemadectin (Desmethoxyamino Moxidectin), TRC

Product Code. 30004240
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004240 5 mg 5mg
30006904 10 mg 10mg
30006277 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004240 Supplier Toronto Research Chemicals Supplier No. TRCK1955355MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112124-81-9
Molecular Formula C36H50O8
Molecular Weight (g/mol) 610.78
Synonym (6R,25S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-6,28-epoxy-23-oxomilbemycin B, [6R,25S(E)]-5-O-Demethyl-28-deoxy-25-(1,3-dimethyl-1-butenyl)-6,28-epoxy-23-oxo-milbemycin B, Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran] Milbemycin B derivative;
IUPAC Name (2S,2a'E,2a1'S,4'E,5R,6S,6'R,8'E,11'R,15'S,17a'R,20'R,20a'R)-2a1',20'-dihydroxy-5,6',8',19'-tetramethyl-6-((E)-4-methylpent-2-en-2-yl)-2a1',5,6,6',7',10',11',14',15',17a',20',20a'-dodecahydro-2'H,17'H-spiro[pyran-2,13'-[11,15]methano[1,5]dioxacyclooctadecino[9,8,7-cd]benzofuran]-4,17'(3H)-dione
SMILES C/C(C[C@@H](C)/C=C/C=C(CO1)/[C@]2(O)[C@@]1([H])[C@H](O)C(C)=C[C@@]2([H])C3=O)=C\C[C@]4([H])C[C@@](O3)([H])C[C@@]5(O[C@H](/C(C)=C/C(C)C)[C@@H](C)C(C5)=O)O4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C36H50O8/c1-20(2)13-24(6)32-25(7)30(37)18-35(44-32)17-28-16-27(43-35)12-11-22(4)14-21(3)9-8-10-26-19-41-33-31(38)23(5)15-29(34(39)42-28)36(26,33)40/h8-11,13,15,20-21,25,27-29,31-33,38,40H,12,14,16-19H2,1-7H3/b9-8+,22-11+,24-13+,26-10+/t21-,25-,27+,28-,29-,31+,32+,33+,35-,36+/m0/s1
Formula Weight 610.35
Percent Purity >95%
Chemical Name or Material 23-Keto Nemadectin (Desmethoxyamino Moxidectin)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.