Learn More
Ibidi 25 Culture-Inserts 4 Well for Self-insertion
Click to view available options
No. of Wells:
4
Unit Size:
Pack of 25
Description
- 25 x 4 well silicone insert with 4 defined cell-free gaps of 500 μm
- suitable for wound healing
- migration assays
- 2D invasion assays
- and co-cultivation of cells
- in a 10 cm transport dish
- sterilized
Specifications
Specifications
| Type | Culture Insert |
| Quantity | 25 |
| Sterility | Sterile |
| No. of Wells | 4 |