missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 06651600 malonate

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
PF 06551600 malonate is a potent and selective JAK3 inhibitor (IC50 = 33.1 μM). Exhibits no significant activity against a panel of other kinases including JAK1, JAK2 and TYK2. Inhibits phosphorylation of STAT5, induced by IL-2, IL-4, IL-7 and IL-15. Suppresses Th1 and Th17 T-cell differentiation and function in vitro. Reduces paw swelling in a rat model of arthritis and reduces disease severity in a rat EAE model.
Specifications
Specifications
| Chemical Name or Material | 1-[(2S,5R)-2-Methyl-5-(7H-pyrrolo[2,3-d]pyrimidin-4-ylamino)-1-piperidinyl]-2-propen-1-one malate |
| CAS | 2140301-97-7 |
| Quantity | 25 mg |
| Molecular Formula | C15H19N5O.C3H4O4 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction