missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ (R)-ZINC 3573
GREENER_CHOICE

Product Code. 18756076
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18756076

Brand: Tocris Bioscience™ 6351/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MRGPRX2 agonist

(R)-ZINC 3573 is a Mas-related G protein-coupled receptor X2 (MRGPRX2) agonist (EC50 = 0.74 μM), which displays selectivity for MRGPRX2 over 350 other GPCRs, including MRGPRX1. (R)-ZINC 3573 induces intracellular calcium release and degranulation in LAD2 mast cells in vitro.

Negative control (S)-ZINC 3573 (Cat. No. 6352) also available.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Molecular Formula C18H21N5
Formula Weight 307.4
Product Type Agonist
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.