Learn More
Abnova™ 25 mM MgCl2
Product Code. 16082565
Click to view available options
Quantity:
1.8 mL
5 x 1.8 mL
Unit Size:
1.8mL
Pack of 5
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 16082565
Brand: Abnova™ undefined
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
25 mM MgCl2 can be used for optimization of magnesium ion concentration in PCR reaction.
Specifications
For Use With (Application) Standard PCR
Quantity 1.8 mL