Learn More
B Braun™ 250 mL Braunoderm™ Preoperation Skin Disinfectant
Product Code. 15813692
Change view
Click to view available options
Quantity:
20/Pk.
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15813692 20/Pk. Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15813692 Supplier B Braun™ Supplier No. 3881105
  • ONLINE EXCLUSIVE PRICE
    € 220.00
    € 212.19 / Pack of 20
    Save € 7.81
    3% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A ready-to-use, quick-acting skin disinfectant for use prior to surgery.
  • Combined from alcohol and PVP-iodine
  • For skin disinfection in case of surgical procedures or aspiration
  • Short exposure time from 15 seconds
  • Long-term active
  • Easy to spot disinfected skin areas
  • Active against bacteria (including MRSA, TbB) and fungal infestation, as well as viruses such as vaccinia and polio
  • DGHM/VAH listed
Specifications
Product Type Skin Disinfectant
Certifications/Compliance DGHM/VAH-listed
Composition Alcohol and PVP-iodine solution
For Use With (Application) Pre-surgical skin disinfection
Quantity 20/Pk.
Volume (Metric) 250 mL