Learn More
2-Bromo-3,3-dimethyl-1-butanol, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-3,3-dimethyl-1-butanol |
| CAS | 21020-18-8 |
| Molecular Formula | C6H13BrO |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C6H13BrO/c1-6(2,3)5(7)4-8/h5,8H,4H2,1-3H3 |
| SMILES | CC(C)(C)C(Br)CO |
| IUPAC Name | 2-bromo-3,3-dimethylbutan-1-ol |
| Molecular Weight (g/mol) | 181.07 |
| Formula Weight | 180.015 |